N-Methyl-4-nitro-N-[2-(4-nitrophenoxy)ethyl]phenylethylamine 97%

Supplier: Ambeed
Danger

Synonyms: N-Methyl-2-(4-nitrophenoxy)-N-[2-(4-nitrophenyl)ethyl]ethanamine

A124477-1G A124477-250MG
77149-354EA 38.11 USD
77149-354 77149-356
N-Methyl-4-nitro-N-[2-(4-nitrophenoxy)ethyl]phenylethylamine 97%
N-Methyl-4-nitro-N-[2-(4-nitrophenoxy)ethyl]phenylethylamine

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃O₅
MW: 345.36 g/mol
MDL Number: MFCD08236712
CAS Number: 115287-37-1
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR