Purmorphamine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(1-Naphthoxy)-6-(4-morpholinoanilino)-9-cyclohexylpurine, 9-Cyclohexyl-N-(4-morpholinophenyl)-2-(naphthalen-1-yloxy)-9H-purin-6-amine

A124253-10MG A124253-1MG A124253-5MG A124253-50MG A124253-100MG A124253-250MG
77149-342EA 67.98 USD
77149-342 77149-344 77149-346 77571-458 77611-060 77685-089
Purmorphamine 98%
Purmorphamine

Precaución: for Research Use Only

Formula: C₃₁H₃₂N₆O₂
MW: 520.637 g/mol
MDL Number: MFCD09037557
CAS Number: 483367-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR