5-Bromo-2,3-difluorophenol 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,4-difluoro-5-hydroxybenzene

A124014-100G A124014-10G A124014-25G A124014-5G
77149-330EA 90 USD
77149-330 77149-332 77149-334 77149-336
5-Bromo-2,3-difluorophenol 95%
5-Bromo-2,3-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
MDL Number: MFCD01631350
CAS Number: 186590-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR