1-(6-Bromo-3-pyridyl)-4-methylpiperazine 95%

Supplier: Ambeed
Warning

A123962-100MG A123962-1G A123962-250MG
77149-318EA 61.8 USD
77149-318 77149-320 77149-322
1-(6-Bromo-3-pyridyl)-4-methylpiperazine 95%
1-(6-Bromo-3-pyridyl)-4-methylpiperazine

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrN₃
MW: 256.14 g/mol
MDL Number: MFCD14702603
CAS Number: 879488-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR