1-Azetidinecarboxylic acid-3-aminophenylmethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-Aminoazetidine, N1-CBZ-protected, 1-Cbz-3-Aminoazetidine

A123257-10G A123257-1G A123257-250MG A123257-25G A123257-5G
77149-306EA 201.88 USD
77149-306 77149-308 77149-310 77149-312 77149-314
1-Azetidinecarboxylic acid-3-aminophenylmethyl ester 98%
1-Azetidinecarboxylic acid-3-aminophenylmethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₂
MW: 206.25 g/mol
MDL Number: MFCD07772068
CAS Number: 112257-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR