4-(2-((2R,3R)-3-(2,5-Difluorophenyl)-3-hydroxy-4-(1H-1,2,4-triazol-1-yl)butan-2-yl)thiazol-4-yl)benzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: Isavuconazole, 4-[2-[(1R,2R)-2-(2,5-Difluorophenyl)-2-hydroxy-1-methyl-3-(1H-1,2,4-triazol-1-yl)propyl]-4-thiazolyl]benzonitrile

A123222-10MG A123222-1MG A123222-25MG A123222-50MG A123222-5MG A123222-100MG A123222-1G A123222-250MG
77149-296EA 19.57 USD
77149-296 77149-298 77149-300 77149-302 77149-304 77566-346 77685-083 77685-084
4-(2-((2R,3R)-3-(2,5-Difluorophenyl)-3-hydroxy-4-(1H-1,2,4-triazol-1-yl)butan-2-yl)thiazol-4-yl)benzonitrile 98%
4-(2-((2R,3R)-3-(2,5-Difluorophenyl)-3-hydroxy-4-(1H-1,2,4-triazol-1-yl)butan-2-yl)thiazol-4-yl)benzonitrile

Precaución: For research use only.

Formula: C₂₂H₁₇F₂N₅OS
MW: 437.47 g/mol
MDL Number: MFCD06407745
CAS Number: 241479-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR