FRAX597 98+%

Supplier: Ambeed
Warning

Synonyms: 6-(2-Chloro-4-(thiazol-5-yl)phenyl)-8-ethyl-2-((4-(4-methylpiperazin-1-yl)phenyl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one, 6-[2-Chloro-4-(5-thiazolyl)phenyl]-8-ethyl-2-[[4-(4-methyl-1-piperazinyl)phenyl]amino]pyrido[2,3-d]pyrimidin-7-(8H)-one

A123005-10MG A123005-1MG A123005-25MG A123005-2MG A123005-5MG
77149-286EA 110.21 USD
77149-286 77149-288 77149-290 77149-292 77149-294
FRAX597 98+%
FRAX597

Precaución: for Research Use Only

Formula: C₂₉H₂₈ClN₇OS
MW: 558.1 g/mol
MDL Number: MFCD25976723
CAS Number: 1286739-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR