LY573636 98%

Supplier: Ambeed
Warning

Synonyms: Tasisulam, N-((5-Bromothiophen-2-yl)sulphonyl)-2,4-dichlorobenzamide, LY 573636

A122315-1MG A122315-25MG A122315-5MG A122315-10MG A122315-50MG
77149-256EA 11.33 USD
77149-256 77149-258 77149-260 77611-044 77685-082
LY573636 98%
LY573636

Precaución: for Research Use Only

Formula: C₁₁H₆BrCl₂NO₃S₂
MW: 415.097 g/mol
CAS Number: 519055-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR