Ethynylferrocene 97%

Supplier: Ambeed
Warning

Synonyms: 1-cyclopenta-2,4-dienyl-(2-ethynyl-1-cyclopenta-2,4-dienyl)iron, Ferrocene, ethynyl-, (2α)-, (2α)-Ethynylferrocene

A122142-10G A122142-1G A122142-250MG A122142-5G
77149-240EA 185 USD
77149-240 77149-242 77149-244 77149-246
Ethynylferrocene 97%
Ethynylferrocene

Precaución: for Research Use Only

Formula: C₁₂H₁₀Fe
MDL Number: MFCD03701501
CAS Number: 1271-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR