Ethylene di(acetate) 98%

Supplier: Ambeed
Warning

Synonyms: Ethane-1,2-diyl diacetate, Ethylene glycol diacetate, Ethylene Glycol Diacetate, 1,2-Diacetoxyethane

A121919-1000G A121919-100G A121919-500G
77149-234EA 21.63 USD
77149-234 77149-236 77149-238
Ethylene di(acetate) 98%
Ethylene di(acetate)

Precaución: for Research Use Only

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
MDL Number: MFCD00008718
CAS Number: 111-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR