1-(2,4-Dihydroxyphenyl)-2-(4-methoxyphenyl)ethanone 95+%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,4-Dihydroxyphenyl 4-Methoxybenzyl ketone, 2',4'-Dihydroxy-2-(4-methoxyphenyl)acetophenone
A121839-1G
A121839-250MG
A121839-5G
77149-228EA
100.94
USD
77149-228
77149-230
77149-232
1-(2,4-Dihydroxyphenyl)-2-(4-methoxyphenyl)ethanone 95+%
1-(2,4-Dihydroxyphenyl)-2-(4-methoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₁₅H₁₄O₄ MW: 258.28 g/mol |
MDL Number:
MFCD00020119 CAS Number: 487-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...