Eupatilin 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3,4-Dimethoxyphenyl)-5,7-dihydroxy-6-methoxy-4H-chromen-4-one, 5,7-Dihydroxy-3',4',6-trimethoxyflavone

A121685-10MG A121685-1MG A121685-25MG A121685-50MG A121685-5MG A121685-1G A121685-250MG A121685-100MG
77149-206EA 33.99 USD
77149-206 77149-208 77149-210 77149-212 77149-214 77683-094 77683-095 77683-093
Eupatilin 98%
Eupatilin

Precaución: For research use only.

Formula: C₁₈H₁₆O₇
MW: 344.3 g/mol
MDL Number: MFCD13194819
CAS Number: 22368-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR