tert-Butyl 1-(hydroxymethyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate 98%

Supplier: Ambeed
Warning

A120521-5G A120521-1G A120521-100MG A120521-250MG
77149-172EA 1087 USD
77149-172 77571-224 77891-859 77891-860
tert-Butyl 1-(hydroxymethyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate 98%
tert-Butyl 1-(hydroxymethyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MDL Number: MFCD18073342
CAS Number: 161152-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR