5-Bromo-2,7-dimethyl-2H-indazole 95%

Supplier: Ambeed
Warning

A120081-1G A120081-250MG A120081-5G
77149-162EA 148.32 USD
77149-162 77149-164 77149-166
5-Bromo-2,7-dimethyl-2H-indazole 95%
5-Bromo-2,7-dimethyl-2H-indazole

Precaución: for Research Use Only

Formula: C₉H₉BrN₂
MDL Number: MFCD12028637
CAS Number: 1146637-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR