1-(2-Bromophenyl)-1H-pyrazole 96%

Supplier: Ambeed
Warning

A120076-1G A120076-250MG A120076-5G
77149-156EA 44.29 USD
77149-156 77149-158 77149-160
1-(2-Bromophenyl)-1H-pyrazole 96%
1-(2-Bromophenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.069 g/mol
MDL Number: MFCD06659065
CAS Number: 87488-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR