Methyl 3-((tert-butoxycarbonyl)amino)bicyclo[1.1.1]pentane-1-carboxylate 95%

Supplier: Ambeed
Warning

A119638-1G A119638-250MG A119638-5G
77149-144EA 142.14 USD
77149-144 77149-146 77149-148
Methyl 3-((tert-butoxycarbonyl)amino)bicyclo[1.1.1]pentane-1-carboxylate 95%
Methyl 3-((tert-butoxycarbonyl)amino)bicyclo[1.1.1]pentane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄
MW: 241.284 g/mol
MDL Number: MFCD24476148
CAS Number: 676371-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR