7-Hydroxychroman-2-one 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A119493-100MG A119493-250MG A119493-1G
77149-136EA 28 USD
77149-136 77149-138 77683-073
7-Hydroxychroman-2-one 95%, Ambeed.Inc

Precaución: For research use only.

Formula: C₉H₈O₃
MDL Number: MFCD00557356
CAS Number: 5631-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR