2,4,6-Trichlorophenyl formate 95%

Supplier: Ambeed
Warning

Synonyms: Formic Acid 2,4,6-Trichlorophenyl Ester, 2,4,6-Trichlorophenyl Formate

A119450-100G A119450-10G A119450-1G A119450-25G A119450-5G
77149-128EA 147 USD
77149-128 77149-130 77149-132 77149-134 77571-158
2,4,6-Trichlorophenyl formate 95%
2,4,6-Trichlorophenyl formate

Precaución: for Research Use Only

Formula: C₇H₃Cl₃O₂
MDL Number: MFCD27996252
CAS Number: 4525-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR