Benzyl-3-hydroxypyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Benzyl 3-hydroxypyrrolidine-1-carboxylate

A119384-10G A119384-1G A119384-5G A119384-25G
77149-122EA 67.98 USD
77149-122 77149-124 77149-126 77611-000
Benzyl-3-hydroxypyrrolidine-1-carboxylate 95%
Benzyl-3-hydroxypyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MW: 221.2500 g/mol
MDL Number: MFCD08061949
CAS Number: 95656-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR