N-(5-(4-(3,3-Dimethylazetidine-1-carbonyl)phenyl)-[1,2,4]triazolo[1,5-a]pyridin-2-yl)cyclopropanecarboxamide 98+%

Supplier: Ambeed
Warning

Synonyms: Solcitinib

A119338-10MG A119338-1MG A119338-50MG A119338-5MG
77149-112EA 49.44 USD
77149-112 77149-114 77149-116 77149-118
N-(5-(4-(3,3-Dimethylazetidine-1-carbonyl)phenyl)-[1,2,4]triazolo[1,5-a]pyridin-2-yl)cyclopropanecarboxamide 98+%
N-(5-(4-(3,3-Dimethylazetidine-1-carbonyl)phenyl)-[1,2,4]triazolo[1,5-a]pyridin-2-yl)cyclopropanecarboxamide

Precaución: for Research Use Only

Formula: C₂₂H₂₃N₅O₂
MW: 389.45 g/mol
MDL Number: MFCD26960569
CAS Number: 1206163-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR