2-Chloro-p-benzoquinone 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chlorocyclohexa-2,5-diene-1,4-dione, 2-Chloro-1,4-benzoquinone

A119076-250MG A119076-5G A119076-25G
77149-096EA 19.57 USD
77149-096 77149-098 77685-048
2-Chloro-p-benzoquinone 95%
2-Chloro-p-benzoquinone

Precaución: for Research Use Only

Formula: C₆H₃ClO₂
MW: 142.54 g/mol
MDL Number: MFCD00075260
CAS Number: 695-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR