2-Bromo-3-chloro-5-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A119045-100G A119045-10G A119045-1G A119045-25G A119045-5G
77149-086EA 119 USD
77149-086 77149-088 77149-090 77149-092 77149-094
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine 97%
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂BrClF₃N
MW: 260.44
MDL Number: MFCD00153072
CAS Number: 75806-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR