(S)-(1,4-Dioxan-2-yl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A119002-10G A119002-250MG A119002-5G A119002-1G
77149-080EA 260.59 USD
77149-080 77149-082 77149-084 77566-194
(S)-(1,4-Dioxan-2-yl)methanamine hydrochloride 97%
(S)-(1,4-Dioxan-2-yl)methanamine hydrochloride

Precaución: For research use only.

Formula: C₅H₁₂ClNO₂
MW: 153.61 g/mol
MDL Number: MFCD27918520
CAS Number: 1523541-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR