6,7-Difluoroquinoline 95%

Supplier: Ambeed
Warning

A119000-1G A119000-250MG
77149-076EA 195 USD
77149-076 77149-078
6,7-Difluoroquinoline 95%
6,7-Difluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅F₂N
MW: 165.14 g/mol
MDL Number: MFCD01939054
CAS Number: 127827-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR