4-Bromo-1-butyl-3-(trifluoromethyl)pyrazole 95%
Supplier: Ambeed
|
Warning
|
A118896-10G
A118896-1G
A118896-250MG
A118896-5G
77149-068EA
125
USD
77149-068
77149-070
77149-072
77149-074
4-Bromo-1-butyl-3-(trifluoromethyl)pyrazole 95%
4-Bromo-1-butyl-3-(trifluoromethyl)pyrazole
Precaución: for Research Use Only
Formula:
C₈H₁₀BrF₃N₂ |
MDL Number:
MFCD24448781 CAS Number: 1437794-58-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...