KU-57788 ≥98%

Supplier: Ambeed
Warning

Synonyms: 8-(Dibenzo[b,d]thiophen-4-yl)-2-morpholino-4H-chromen-4-one, NU-7441

A118825-10MG A118825-1MG A118825-5MG
77149-052EA 35.02 USD
77149-052 77149-054 77149-056
KU-57788 ≥98%
KU-57788

Precaución: for Research Use Only

Formula: C₂₅H₁₉NO₃S
MW: 413.5 g/mol
MDL Number: MFCD11983069
CAS Number: 503468-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR