5-Bromo-2-iodobenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (5-Bromo-2-iodophenyl)methanol

A118819-10G A118819-1G A118819-250MG A118819-25G A118819-5G
77149-042EA 398.61 USD
77149-042 77149-044 77149-046 77149-048 77149-050
5-Bromo-2-iodobenzyl alcohol 97%
5-Bromo-2-iodobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrIO
MW: 312.93 g/mol
MDL Number: MFCD09745850
CAS Number: 199786-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR