TAK-733 ≥98%

Supplier: Ambeed
Warning

Synonyms: (R)-3-(2,3-Dihydroxypropyl)-6-fluoro-5-((2-fluoro-4-iodophenyl)amino)-8-methylpyrido[2,3-d]pyrimidine-4,7(3H,8H)-dione

A118745-10MG A118745-2MG A118745-5MG
77149-036EA 241.02 USD
77149-036 77149-038 77149-040
TAK-733 ≥98%
TAK-733

Precaución: for Research Use Only

Formula: C₁₇H₁₅F₂IN₄O₄
MW: 504.23 g/mol
MDL Number: MFCD24386349
CAS Number: 1035555-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR