Ethyl-2-amino-2-methylpropanoate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: H-Aib-OEt.HCl, H-Aib-OEt·HCl

A118334-10G A118334-1G A118334-250MG A118334-25G A118334-5G
77149-020EA 20.6 USD
77149-020 77149-022 77149-024 77149-026 77149-028
Ethyl-2-amino-2-methylpropanoate hydrochloride 95%
Ethyl-2-amino-2-methylpropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂
MW: 167.633 g/mol
MDL Number: MFCD06809830
CAS Number: 17288-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR