tert-Butyl (2-(methylamino)ethyl)carbamate 96%

Supplier: Ambeed
Warning

A118241-100G A118241-10G A118241-1G A118241-25G A118241-5G
77149-006EA 147 USD
77149-006 77149-008 77149-010 77149-012 77149-014
tert-Butyl (2-(methylamino)ethyl)carbamate 96%
tert-Butyl (2-(methylamino)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₂
MDL Number: MFCD06808585
CAS Number: 122734-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR