1-Bromo-2,4-dichloro-3-methylbenzene 98%

Supplier: Ambeed
Warning

A118196-1G A118196-5G
77149-002EA 27 USD
77149-002 77149-004
1-Bromo-2,4-dichloro-3-methylbenzene 98%
1-Bromo-2,4-dichloro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MDL Number: MFCD09878179
CAS Number: 127049-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR