Fmoc-N(Hmb)-Gly-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-(Hmb)Gly-OH

A117862-100MG A117862-1G A117862-5G A117862-250MG
77148-992EA 6 USD
77148-992 77148-994 77566-158 77685-043
Fmoc-N(Hmb)-Gly-OH 97%
Fmoc-N(Hmb)-Gly-OH

Precaución: For research use only.

Formula: C₂₅H₂₃NO₆
MW: 433.46 g/mol
MDL Number: MFCD03093500
CAS Number: 148515-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR