Fmoc-β-Ala-OPFP 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-β-Ala-OPfp

A117741-10G A117741-1G A117741-250MG A117741-5G
77148-984EA 380.07 USD
77148-984 77148-986 77148-988 77148-990
Fmoc-β-Ala-OPFP 98%
Fmoc-beta-Ala-OPFP

Precaución: for Research Use Only

Formula: C₂₄H₁₆F₅NO₄
MW: 477.387 g/mol
MDL Number: MFCD00237655
CAS Number: 149303-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR