3-(8-Chloroimidazo[1,5-a]pyrazin-3-yl)cyclobutanone 95%

Supplier: Ambeed
Warning

A117282-1G A117282-250MG
77148-976EA 498 USD
77148-976 77148-978
3-(8-Chloroimidazo[1,5-a]pyrazin-3-yl)cyclobutanone 95%
3-(8-Chloroimidazo[1,5-a]pyrazin-3-yl)cyclobutanone

Precaución: for Research Use Only

Formula: C₁₀H₈ClN₃O
MW: 221.64 g/mol
MDL Number: MFCD23106397
CAS Number: 936901-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR