3,5-Dichloro-6-ethylpyrazine-2-carboxylic acid 96%

Supplier: Ambeed
Warning

A117008-100MG A117008-1G A117008-250MG
77148-970EA 86 USD
77148-970 77148-972 77148-974
3,5-Dichloro-6-ethylpyrazine-2-carboxylic acid 96%
3,5-Dichloro-6-ethylpyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₂
MDL Number: MFCD28506305
CAS Number: 1883347-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR