(4-Chloro-3-nitrophenyl)(4-fluorophenyl)methanone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-4'-fluoro-3-nitrobenzophenone
A115835-100G
A115835-10G
A115835-1G
A115835-250MG
A115835-25G
A115835-5G
77148-920EA
634.48
USD
77148-920
77148-922
77148-924
77148-926
77148-928
77148-930
(4-Chloro-3-nitrophenyl)(4-fluorophenyl)methanone 97%
(4-Chloro-3-nitrophenyl)(4-fluorophenyl)methanone
Precaución: for Research Use Only
Formula:
C₁₃H₇ClFNO₃ MW: 279.66 g/mol |
MDL Number:
MFCD03840494 CAS Number: 31431-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...