4-Hydroxy-3,6-dimethyl-2(1H)-pyridinone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-3,6-dimethylpyridin-2(1H)-one

A115823-100MG A115823-1G A115823-250MG
77148-914EA 56.65 USD
77148-914 77148-916 77148-918
4-Hydroxy-3,6-dimethyl-2(1H)-pyridinone 97%
4-Hydroxy-3,6-dimethyl-2(1H)-pyridinone

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.152 g/mol
MDL Number: MFCD11113422
CAS Number: 4664-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR