GDC-0068 ≥98%

Supplier: Ambeed
Warning

Synonyms: Ipatasertib, (S)-2-(4-chlorophenyl)-1-(4-((5R,7R)-7-hydroxy-5-methyl-6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-yl)piperazin-1-yl)-3-(isopropylamino)propan-1-one

A115731-10MG A115731-5MG
77148-910EA 74.16 USD
77148-910 77148-912
GDC-0068 ≥98%
GDC-0068

Precaución: for Research Use Only

Formula: C₂₄H₃₂ClN₅O₂
MW: 458.003 g/mol
MDL Number: MFCD22124514
CAS Number: 1001264-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR