4-(Aminomethyl)tetrahydropyran 97%

Supplier: Ambeed
Danger

Synonyms: 4-(Aminomethyl)tetrahydro-2H-pyran, 4-Aminomethyltetrahydropyran, Tetrahydro-2H-pyran-4-ylmethanamine, (Tetrahydro-2H-pyran-4-yl)methanamine

A115642-1G A115642-25G A115642-5G A115642-10G
77148-904EA 5 USD
77148-904 77148-906 77148-908 77570-950
4-(Aminomethyl)tetrahydropyran 97%
4-(Aminomethyl)tetrahydropyran

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MDL Number: MFCD02179435
CAS Number: 130290-79-8
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR