Methyl 4-bromo-5-chlorothiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A115306-1G A115306-250MG A115306-25G
77148-886EA 6 USD
77148-886 77148-888 77148-890
Methyl 4-bromo-5-chlorothiophene-2-carboxylate 97%
Methyl 4-bromo-5-chlorothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄BrClO₂S
MDL Number: MFCD28359275
CAS Number: 1047630-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR