AdipoRon 98%

Supplier: Ambeed
Warning

A115127-10MG A115127-1MG A115127-50MG A115127-5MG A115127-100MG A115127-250MG
77148-878EA 30 USD
77148-878 77148-880 77148-882 77148-884 77685-019 77685-020
AdipoRon 98%
AdipoRon

Precaución: for Research Use Only

Formula: C₂₇H₂₈N₂O₃
MDL Number: MFCD08271060
CAS Number: 924416-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR