N-Cbz-L-serine 97%

Supplier: Ambeed
Warning

Synonyms: N-Carbobenzyloxy-L-serine, Z-Ser-OH, N-Cbz-L-serine

A114416-1000G A114416-100G A114416-10G A114416-25G A114416-500G A114416-5G
77148-848EA 309 USD
77148-848 77148-850 77148-852 77148-854 77148-856 77148-858
N-Cbz-L-serine 97%
N-Cbz-L-serine

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₅
MW: 239.2 g/mol
MDL Number: MFCD00002662
CAS Number: 1145-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR