2-(2,4-Dihydroxyphenyl)-5,7-dihydroxy-3,8-bis(3-methylbut-2-en-1-yl)-4H-chromen-4-one 98%

Supplier: Ambeed
Warning

A114413-10MG A114413-1MG A114413-5MG
77148-842EA 323.42 USD
77148-842 77148-844 77148-846
2-(2,4-Dihydroxyphenyl)-5,7-dihydroxy-3,8-bis(3-methylbut-2-en-1-yl)-4H-chromen-4-one 98%
2-(2,4-Dihydroxyphenyl)-5,7-dihydroxy-3,8-bis(3-methylbut-2-en-1-yl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₅H₂₆O₆
MW: 422.47 g/mol
MDL Number: MFCD20274692
CAS Number: 62949-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR