3-(1H-Pyrazol-1-yl)propane-1,2-diol 96%

Supplier: Ambeed
Warning

A114151-1G A114151-250MG
77148-840EA 88 USD
77148-840 77900-616
3-(1H-Pyrazol-1-yl)propane-1,2-diol 96%
3-(1H-Pyrazol-1-yl)propane-1,2-diol

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O₂
MDL Number: MFCD11846459
CAS Number: 98484-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR