3-Chloro-2-fluoro-4-iodopyridine 95%

Supplier: Ambeed
Warning

A113996-1G A113996-250MG
77148-836EA 18 USD
77148-836 77148-838
3-Chloro-2-fluoro-4-iodopyridine 95%
3-Chloro-2-fluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂ClFIN
MDL Number: MFCD16610366
CAS Number: 796851-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR