3-(4-Bromophenoxy)propanenitrile 97%

Supplier: Ambeed
Warning

A113991-1G A113991-250MG
77148-832EA 43.26 USD
77148-832 77148-834
3-(4-Bromophenoxy)propanenitrile 97%
3-(4-Bromophenoxy)propanenitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MDL Number: MFCD00634314
CAS Number: 118449-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR