4,6-Diaminoresorcinol dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dihydroxy-1,3-phenylenediamine Dihydrochloride, 4,6-Diaminobenzene-1,3-diol dihydrochloride

A113107-1G A113107-250MG A113107-25G A113107-5G
77148-802EA 80.34 USD
77148-802 77148-804 77148-806 77148-808 77148-810
4,6-Diaminoresorcinol dihydrochloride 97%
4,6-Diaminoresorcinol dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MDL Number: MFCD00143239
CAS Number: 16523-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR