6-Bromo-3-iodoquinoline 95%

Supplier: Ambeed
Warning

A112821-1G A112821-250MG
77148-778EA 136.99 USD
77148-778 77148-780
6-Bromo-3-iodoquinoline 95%
6-Bromo-3-iodoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrIN
MW: 333.95 g/mol
MDL Number: MFCD22690575
CAS Number: 1416440-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR