Boc-4-hydrazinobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(2-(tert-Butoxycarbonyl)hydrazinyl)benzoic acid

A112602-1G A112602-25G A112602-5G
77148-772EA 27.81 USD
77148-772 77148-774 77148-776
Boc-4-hydrazinobenzoic acid 97%
Boc-4-hydrazinobenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₄
MW: 252.3 g/mol
MDL Number: MFCD06656437
CAS Number: 96736-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR