(R)-(3-(3-Fluoro-4-(6-(2-methyl-2H-tetrazol-5-yl)pyridin-3-yl)phenyl)-2-oxooxazolidin-5-yl)methyl dihydrogen phosphate 98%

Supplier: Ambeed
Danger

Synonyms: Tedizolid phosphate

A112409-100MG A112409-10MG A112409-1G A112409-250MG A112409-50MG A112409-5G A112409-5MG
77148-760EA 12.36 USD
77148-760 77148-762 77148-764 77148-766 77148-768 77148-770 77592-196
(R)-(3-(3-Fluoro-4-(6-(2-methyl-2H-tetrazol-5-yl)pyridin-3-yl)phenyl)-2-oxooxazolidin-5-yl)methyl dihydrogen phosphate 98%
(R)-(3-(3-Fluoro-4-(6-(2-methyl-2H-tetrazol-5-yl)pyridin-3-yl)phenyl)-2-oxooxazolidin-5-yl)methyl dihydrogen phosphate

Precaución: for Research Use Only

Formula: C₁₇H₁₆FN₆O₆P
MW: 450.32 g/mol
MDL Number: MFCD28098176
CAS Number: 856867-55-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR